Product Details
- SNP ID
-
rs199926769
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:144074321 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTGGGCTCTTCTCCCTGTTCTA[C/T]ATCTTCATTCTGTTGGGGAACGGGA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR2A25
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs59319753] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR2A25
- Gene Name
- olfactory receptor family 2 subfamily A member 25
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004488.1 |
102 |
Silent Mutation |
TAC,TAT |
Y34Y |
NP_001004488.1 |
View Full Product Details