Product Details

SNP ID
rs200605076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100160229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGCCACAGCTGTCTGCAGCCGG[C/T]CCTGGCCGTATTTCTCTATCCGTGC
Phenotype
MIM: 608235
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf43 PubMed Links

Gene Details

Gene
C7orf43
Gene Name
chromosome 7 open reading frame 43
There are no transcripts associated with this gene.

Gene
GAL3ST4
Gene Name
galactose-3-O-sulfotransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024637.4 1553 Missense Mutation GAC,GGC D387G NP_078913.3
Gene
GPC2
Gene Name
glypican 2
There are no transcripts associated with this gene.

Gene
LAMTOR4
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 4
There are no transcripts associated with this gene.

Gene
MIR4658
Gene Name
microRNA 4658
There are no transcripts associated with this gene.

View Full Product Details