Product Details

SNP ID
rs200794657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:4799394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCCCCCTAGAGAGGGGGCGTG[C/T]GGAAATGGATCTTCTTGGCTGTTTC
Phenotype
MIM: 613653 MIM: 611491
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP5Z1 PubMed Links

Gene Details

Gene
AP5Z1
Gene Name
adaptor related protein complex 5 zeta 1 subunit
There are no transcripts associated with this gene.

Gene
RADIL
Gene Name
Rap associating with DIL domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018059.4 3385 Missense Mutation CAC,CGC H1071R NP_060529.4

View Full Product Details