Product Details

SNP ID
rs200813675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99419402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCGTTGCAGGGCCGCATCAT[C/T]GAGTGCACACACTGTGGCTGTCGTG
Phenotype
MIM: 603477 MIM: 614774
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5J2-PTCD1 PubMed Links

Gene Details

Gene
ATP5J2-PTCD1
Gene Name
ATP5J2-PTCD1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198879.1 2842 UTR 3 NP_001185808.1
Gene
BUD31
Gene Name
BUD31 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003910.3 2842 Silent Mutation ATC,ATT I132I NP_003901.2
XM_005250670.4 2842 Intron XP_005250727.1
XM_005250671.4 2842 Intron XP_005250728.1
XM_005250674.3 2842 Silent Mutation ATC,ATT I132I XP_005250731.1
XM_017012760.1 2842 Intron XP_016868249.1
XM_017012761.1 2842 Intron XP_016868250.1
XM_017012762.1 2842 Silent Mutation ATC,ATT I132I XP_016868251.1
Gene
PTCD1
Gene Name
pentatricopeptide repeat domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015545.3 2842 UTR 3 NP_056360.2

View Full Product Details