Product Details

SNP ID
rs201061684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149436652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCTTCCGTGGACTCCTCCAGC[A/G]TGTTCTCTGGCTCCTGCATGTCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF777 PubMed Links

Gene Details

Gene
ZNF777
Gene Name
zinc finger protein 777
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015694.2 1435 Missense Mutation ACG,ATG T421M NP_056509.2
XM_005249980.3 1435 Missense Mutation ACG,ATG T465M XP_005250037.1
XM_011516055.2 1435 Missense Mutation ACG,ATG T421M XP_011514357.1

View Full Product Details