Product Details

SNP ID
rs200039404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73840110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGCACCGACTATCAGCACCGC[G/T]TCGAAGGTCCCTGTGTGTGTGTGGG
Phenotype
MIM: 602909 MIM: 612546
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLDN4 PubMed Links

Gene Details

Gene
CLDN4
Gene Name
claudin 4
There are no transcripts associated with this gene.

Gene
WBSCR27
Gene Name
Williams Beuren syndrome chromosome region 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152559.2 441 Missense Mutation GAA,GAC E133D NP_689772.2
XM_017011777.1 441 Missense Mutation GAA,GAC E133D XP_016867266.1
XM_017011778.1 441 Missense Mutation GAA,GAC E133D XP_016867267.1

View Full Product Details