Product Details

SNP ID
rs200491602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140521990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAAGGCCTCCCGCTGCAGGGTTC[A/T]CTCCTCACGCTCGCCCGACGTCAGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DENND2A PubMed Links

Gene Details

Gene
DENND2A
Gene Name
DENN domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318052.1 3278 Nonsense Mutation AGA,TGA R926* NP_001304981.1
NM_001318053.1 3278 Intron NP_001304982.1
NM_015689.4 3278 Nonsense Mutation AGA,TGA R926* NP_056504.3
XM_005249977.3 3278 Nonsense Mutation AGA,TGA R926* XP_005250034.1
XM_005249979.4 3278 Intron XP_005250036.1
XM_011516052.2 3278 Nonsense Mutation AGA,TGA R926* XP_011514354.1
XM_011516053.2 3278 Nonsense Mutation AGA,TGA R926* XP_011514355.1
XM_011516054.2 3278 Nonsense Mutation AGA,TGA R925* XP_011514356.1
XM_017011989.1 3278 Nonsense Mutation AGA,TGA R926* XP_016867478.1

View Full Product Details