Product Details

SNP ID
rs200795996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100805302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGTAGTGAGGCTGCCGCTGGT[C/G]CAGGAGAGGGTGTGAGGCCCTAGGG
Phenotype
MIM: 600011 MIM: 602372
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EPHB4 PubMed Links

Gene Details

Gene
EPHB4
Gene Name
EPH receptor B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004444.4 2846 Missense Mutation CAC,GAC H900D NP_004435.3
XM_017011816.1 2846 Missense Mutation CAC,GAC H918D XP_016867305.1
Gene
ZAN
Gene Name
zonadhesin (gene/pseudogene)
There are no transcripts associated with this gene.

View Full Product Details