Product Details

SNP ID
rs199930483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27108368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGGGCTTGGAGAAGGGCGGGGG[A/C]GACTGGGGCTCATACGGGACGCTGT
Phenotype
MIM: 604685 MIM: 142954
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HOTAIRM1 PubMed Links

Gene Details

Gene
HOTAIRM1
Gene Name
HOXA transcript antisense RNA, myeloid-specific 1
There are no transcripts associated with this gene.

Gene
HOXA2
Gene Name
homeobox A2
There are no transcripts associated with this gene.

Gene
HOXA3
Gene Name
homeobox A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030661.4 1782 Silent Mutation TCG,TCT S293S NP_109377.1
NM_153631.2 1782 Silent Mutation TCG,TCT S293S NP_705895.1
XM_005249730.2 1782 Silent Mutation TCG,TCT S293S XP_005249787.1
XM_005249731.2 1782 Silent Mutation TCG,TCT S293S XP_005249788.1
XM_005249732.3 1782 Silent Mutation TCG,TCT S293S XP_005249789.1
XM_006715715.2 1782 Silent Mutation TCG,TCT S293S XP_006715778.1
XM_011515343.2 1782 Silent Mutation TCG,TCT S293S XP_011513645.1

View Full Product Details