Product Details

SNP ID
rs200419920
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149432091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACTTCTCGCTGAAGCTCTTCTC[A/G]CACTCGGGGCACTTGAAGGGCCGCT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF777 PubMed Links

Gene Details

Gene
ZNF777
Gene Name
zinc finger protein 777
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015694.2 2354 Silent Mutation TGC,TGT C727C NP_056509.2
XM_005249980.3 2354 Silent Mutation TGC,TGT C771C XP_005250037.1
XM_011516055.2 2354 Silent Mutation TGC,TGT C727C XP_011514357.1

View Full Product Details