Product Details

SNP ID
rs200985152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143478596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTCCCTCATCTCCTTCCTCATT[C/T]TTTATGCTCTGTCCTTTCTGTCCCT
Phenotype
MIM: 613965
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPHA1-AS1 PubMed Links

Gene Details

Gene
EPHA1-AS1
Gene Name
EPHA1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TAS2R41
Gene Name
taste 2 receptor member 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176883.2 724 Missense Mutation CTT,TTT L242F NP_795364.2

View Full Product Details