Product Details

SNP ID
rs199896416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39951678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAAGTCCCCCAGCCCGGCAGGAG[G/T]TGGCAGCAGCCCCTATTCTCGGCGG
Phenotype
MIM: 603309
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDK13 PubMed Links

Gene Details

Gene
CDK13
Gene Name
cyclin dependent kinase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003718.4 2178 Missense Mutation GGT,GTT G346V NP_003709.3
NM_031267.3 2178 Missense Mutation GGT,GTT G346V NP_112557.2
XM_011515597.2 2178 Missense Mutation GGT,GTT G346V XP_011513899.1
XM_017012750.1 2178 Missense Mutation GGT,GTT G346V XP_016868239.1
XM_017012751.1 2178 Missense Mutation GGT,GTT G346V XP_016868240.1

View Full Product Details