Product Details

SNP ID
rs200306735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:138802936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCACAAGCAGCCCGGTGCTGCC[A/G]CACAGGAGTGGACGAGTACGGCTGG
Phenotype
MIM: 605239
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0A4 PubMed Links

Gene Details

Gene
ATP6V0A4
Gene Name
ATPase H+ transporting V0 subunit a4
There are no transcripts associated with this gene.

Gene
TMEM213
Gene Name
transmembrane protein 213
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085429.1 302 Missense Mutation CAC,CGC H64R NP_001078898.1
XM_005250167.3 302 Missense Mutation CAC,CGC H114R XP_005250224.1

View Full Product Details