Product Details

SNP ID
rs200647083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100463387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCAGCTGCACATCTGCCAGCC[A/G]CTCTGCAGGGCAGAGACCATGAGCG
Phenotype
MIM: 611914
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf61 PubMed Links

Gene Details

Gene
C7orf61
Gene Name
chromosome 7 open reading frame 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004323.1 609 Silent Mutation AGC,AGT S121S NP_001004323.1
Gene
TSC22D4
Gene Name
TSC22 domain family member 4
There are no transcripts associated with this gene.

View Full Product Details