Product Details

SNP ID
rs200213225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100431019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAAGTGGTGTCTCCCCTTCCAT[C/T]TGCTCTGCAGGGTCCCTCAGGCTCC
Phenotype
MIM: 611478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEPCE PubMed Links

Gene Details

Gene
MEPCE
Gene Name
methylphosphate capping enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194990.1 656 UTR 5 NP_001181919.1
NM_001194991.1 656 Intron NP_001181920.1
NM_001194992.1 656 Intron NP_001181921.1
NM_019606.5 656 Missense Mutation TCT,TTT S334F NP_062552.2
XM_011516410.2 656 Intron XP_011514712.1
Gene
PPP1R35
Gene Name
protein phosphatase 1 regulatory subunit 35
There are no transcripts associated with this gene.

Gene
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
There are no transcripts associated with this gene.

View Full Product Details