Product Details

SNP ID
rs200478538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144074728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTACCACTGTCCTTCTGTGGAC[C/G]CCAGAAACTTAATCACTTTTTCTGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR2A25 PubMed Links

Gene Details

Gene
OR2A25
Gene Name
olfactory receptor family 2 subfamily A member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004488.1 509 Missense Mutation CCC,CGC P170R NP_001004488.1

View Full Product Details