Product Details

SNP ID
rs200497385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:142252530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACACAGCATATTTTCCGTGATG[A/T]TGTAGGTTTTATAGGCATCGCGACA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MOXD2P PubMed Links

Gene Details

Gene
MOXD2P
Gene Name
monooxygenase, DBH-like 2, pseudogene
There are no transcripts associated with this gene.

Gene
PRSS58
Gene Name
protease, serine 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001317.4 627 Missense Mutation AAC,ATC N173I NP_001001317.1

View Full Product Details