Product Details

SNP ID
rs200648639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:16861387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAATGAGATCACATACATACCTA[C/G]AAACATGATTCTAGGCACATATTGC
Phenotype
MIM: 609482
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AGR3 PubMed Links

Gene Details

Gene
AGR3
Gene Name
anterior gradient 3, protein disulphide isomerase family member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176813.4 416 Missense Mutation CTA,GTA L122V NP_789783.1
XM_005249627.2 416 Missense Mutation CTA,GTA L122V XP_005249684.1
XM_011515152.2 416 Missense Mutation CTA,GTA L122V XP_011513454.1

View Full Product Details