Product Details
- SNP ID
-
rs200913289
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:1236099 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCTCCGACAGCGACAGCGGCGGC[A/G]GCGGCCTGTCTCCGGAGCCGCCCGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UNCX
PubMed Links
Gene Details
- Gene
- UNCX
- Gene Name
- UNC homeobox
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080461.1 |
718 |
Missense Mutation |
AGC,GGC |
S240G |
NP_001073930.1 |
View Full Product Details