Product Details

SNP ID
rs200385286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:142871922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCTTTGTCCAAATCCTCAGAGC[C/T]CCGGGTGTGGAAGGCCTGTGCGTAG
Phenotype
MIM: 602757 MIM: 606680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPHB6 PubMed Links

Gene Details

Gene
EPHB6
Gene Name
EPH receptor B6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280794.2 2192 Intron NP_001267723.2
NM_001280795.2 2192 Intron NP_001267724.2
NM_004445.5 2192 Intron NP_004436.4
XM_006715881.2 2192 Intron XP_006715944.1
XM_011515879.1 2192 Intron XP_011514181.1
XM_011515880.1 2192 Intron XP_011514182.1
XM_011515881.2 2192 Intron XP_011514183.1
XM_011515882.2 2192 Intron XP_011514184.1
Gene
TRPV6
Gene Name
transient receptor potential cation channel subfamily V member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018646.5 2192 Missense Mutation AGC,GGC S695G NP_061116.5

View Full Product Details