Product Details

SNP ID
rs200556919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:124746960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTACAGGCTTTCTCTGCTTTGCG[C/G]ATTTTCCTCGCAGTCACTAGAGAGC
Phenotype
MIM: 602583
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPR37 PubMed Links

Gene Details

Gene
GPR37
Gene Name
G protein-coupled receptor 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005302.3 2456 Missense Mutation ATC,ATG I469M NP_005293.1

View Full Product Details