Product Details

SNP ID
rs199976092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151239125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTAGGTGTTGCGCACAACCAGC[A/G]ACTGCTCCAGCTCCTGCCTGCGCTG
Phenotype
MIM: 608037 MIM: 615245 MIM: 601737
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHPF2 PubMed Links

Gene Details

Gene
CHPF2
Gene Name
chondroitin polymerizing factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284295.1 1520 Intron NP_001271224.1
NM_019015.2 1520 Intron NP_061888.1
Gene
MIR671
Gene Name
microRNA 671
There are no transcripts associated with this gene.

Gene
SMARCD3
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003801.1 1520 Missense Mutation TCG,TTG S477L NP_001003801.1
NM_001003802.1 1520 Missense Mutation TCG,TTG S464L NP_001003802.1
NM_003078.3 1520 Missense Mutation TCG,TTG S464L NP_003069.2
XM_011516521.2 1520 Missense Mutation TCG,TTG S375L XP_011514823.1
XM_017012556.1 1520 Missense Mutation TCG,TTG S312L XP_016868045.1

View Full Product Details