Product Details

SNP ID
rs200293184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144095554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCCTGGCCTCAACTTGCTGGAT[A/G]TTTAGCTTTCTCTTGGCTCTGGTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2A12 PubMed Links

Gene Details

Gene
OR2A12
Gene Name
olfactory receptor family 2 subfamily A member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004135.1 447 Missense Mutation ATA,ATG I149M NP_001004135.1

View Full Product Details