Product Details

SNP ID
rs200635965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75988282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGTTGTTGATGAGGATGCTCTCC[C/T]GGATGGTCAGGGTGCAGTAGTACCA
Phenotype
MIM: 124015 MIM: 616695 MIM: 616550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POR PubMed Links

Gene Details

Gene
POR
Gene Name
cytochrome p450 oxidoreductase
There are no transcripts associated with this gene.

Gene
STYXL1
Gene Name
serine/threonine/tyrosine interacting-like 1
There are no transcripts associated with this gene.

Gene
TMEM120A
Gene Name
transmembrane protein 120A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317803.1 637 Missense Mutation NP_001304732.1
NM_031925.2 637 Missense Mutation NP_114131.1

View Full Product Details