Product Details

SNP ID
rs200975298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:31339089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTCTTCCCTGTCTTCCTCCTC[C/T]TCTTCTTTCTCGGTTTCTTCTCCAG
Phenotype
MIM: 611513
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEUROD6 PubMed Links

Gene Details

Gene
NEUROD6
Gene Name
neuronal differentiation 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022728.3 533 Silent Mutation GAA,GAG E60E NP_073565.2

View Full Product Details