Product Details

SNP ID
rs201056403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99968187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAGGATATTTTTGCTGTATTTC[A/G]GGTATTTCCGCAGAGTCGCAGGGCA
Phenotype
MIM: 194460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AZGP1 PubMed Links

Gene Details

Gene
AZGP1
Gene Name
alpha-2-glycoprotein 1, zinc-binding
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185.3 Intron NP_001176.1

View Full Product Details