Product Details

SNP ID
rs200072889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:142871948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGTGTGGAAGGCCTGTGCGTAG[C/T]GTTGGATCCGCTGCCGGTTGAGATC
Phenotype
MIM: 602757 MIM: 606680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPHB6 PubMed Links

Gene Details

Gene
EPHB6
Gene Name
EPH receptor B6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280794.2 2166 Intron NP_001267723.2
NM_001280795.2 2166 Intron NP_001267724.2
NM_004445.5 2166 Intron NP_004436.4
XM_006715881.2 2166 Intron XP_006715944.1
XM_011515879.1 2166 Intron XP_011514181.1
XM_011515880.1 2166 Intron XP_011514182.1
XM_011515881.2 2166 Intron XP_011514183.1
XM_011515882.2 2166 Intron XP_011514184.1
Gene
TRPV6
Gene Name
transient receptor potential cation channel subfamily V member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018646.5 2166 Missense Mutation CAC,CGC H686R NP_061116.5

View Full Product Details