Product Details

SNP ID
rs199906112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:4781711 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGGCCTGGGACCACACGCAGA[A/C]CAGCCGGCAGCTGAGCCTGGTGGCC
Phenotype
MIM: 613653
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AP5Z1 PubMed Links

Gene Details

Gene
AP5Z1
Gene Name
adaptor related protein complex 5 zeta 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014855.2 408 Missense Mutation AAC,ACC N108T NP_055670.1
XM_017012864.1 408 Intron XP_016868353.1
Gene
MIR4656
Gene Name
microRNA 4656
There are no transcripts associated with this gene.

View Full Product Details