Product Details

SNP ID
rs200056012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19145087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTAGGTGATCACCCTTTTCCTCTT[G/T]GGGCGGCCTAATAGGGAGACACCTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FERD3L PubMed Links

Gene Details

Gene
FERD3L
Gene Name
Fer3-like bHLH transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152898.2 335 Silent Mutation CCA,CCC P92P NP_690862.1

View Full Product Details