Product Details

SNP ID
rs201896123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100466986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACGGAGGGCCCATTGGGCGCAGG[A/G]GGCCCAAGCCGTGGGACCCCCGAGG
Phenotype
MIM: 611914
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf61 PubMed Links

Gene Details

Gene
C7orf61
Gene Name
chromosome 7 open reading frame 61
There are no transcripts associated with this gene.

Gene
TSC22D4
Gene Name
TSC22 domain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303043.1 1916 Silent Mutation CCC,CCT P387P NP_001289972.1
NM_030935.4 1916 Silent Mutation CCC,CCT P387P NP_112197.1

View Full Product Details