Product Details

SNP ID
rs202239956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75881981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGTGATCTTCGCCATCTTCTCC[A/G]CTATCATCTTCCTGTCATTCGAGGC
Phenotype
MIM: 615203
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHBDD2 PubMed Links

Gene Details

Gene
RHBDD2
Gene Name
rhomboid domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040456.1 374 Missense Mutation ACT,GCT T111A NP_001035546.1
NM_001040457.1 374 UTR 5 NP_001035547.1
XM_005250511.4 374 UTR 5 XP_005250568.1

View Full Product Details