Product Details

SNP ID
rs201803639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144074809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGATACCCACATTAATGAGGTAA[A/T]GGTTTTGGCAGGGGCAGTGTCTGTG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2A25 PubMed Links

Gene Details

Gene
OR2A25
Gene Name
olfactory receptor family 2 subfamily A member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004488.1 590 Missense Mutation AAG,ATG K197M NP_001004488.1

View Full Product Details