Product Details
- SNP ID
-
rs201972241
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:87934479 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACGGCAGCACCATGCAGGCGGCAG[G/T]GGCTGTGTCCGTGCCCTTCTTGCTG
- Phenotype
-
MIM: 603709
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADAM22
PubMed Links
Gene Details
- Gene
- ADAM22
- Gene Name
- ADAM metallopeptidase domain 22
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001324417.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
NP_001311346.1 |
NM_001324418.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
NP_001311347.1 |
NM_001324419.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
NP_001311348.1 |
NM_001324420.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
NP_001311349.1 |
NM_001324421.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
NP_001311350.1 |
NM_004194.4 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
NP_004185.1 |
NM_016351.5 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
NP_057435.2 |
NM_021721.4 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
NP_068367.1 |
NM_021722.5 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
NP_068368.2 |
NM_021723.4 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
NP_068369.1 |
XM_005250445.3 |
229 |
Intron |
|
|
XP_005250502.1 |
XM_006716028.3 |
229 |
Intron |
|
|
XP_006716091.1 |
XM_006716029.3 |
229 |
Intron |
|
|
XP_006716092.1 |
XM_011516318.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_011514620.1 |
XM_011516319.2 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_011514621.1 |
XM_011516320.2 |
229 |
Intron |
|
|
XP_011514622.1 |
XM_011516321.2 |
229 |
Intron |
|
|
XP_011514623.1 |
XM_011516322.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_011514624.1 |
XM_011516323.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_011514625.1 |
XM_011516324.1 |
229 |
Intron |
|
|
XP_011514626.1 |
XM_017012329.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_016867818.1 |
XM_017012330.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_016867819.1 |
XM_017012331.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_016867820.1 |
XM_017012332.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_016867821.1 |
XM_017012333.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_016867822.1 |
XM_017012334.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_016867823.1 |
XM_017012335.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_016867824.1 |
XM_017012336.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_016867825.1 |
XM_017012337.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_016867826.1 |
XM_017012338.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_016867827.1 |
XM_017012339.1 |
229 |
Missense Mutation |
GGG,GTG |
G5V |
XP_016867828.1 |
XM_017012340.1 |
229 |
Intron |
|
|
XP_016867829.1 |
XM_017012341.1 |
229 |
Intron |
|
|
XP_016867830.1 |
View Full Product Details