Product Details

SNP ID
rs201990061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150740903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAGAATGGGAGGATTCCAGAGG[A/G]GCAAATATGGAACTATGGCTGAAGG
Phenotype
MIM: 608086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIMAP1-GIMAP5 PubMed Links

Gene Details

Gene
GIMAP1-GIMAP5
Gene Name
GIMAP1-GIMAP5 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199577.1 412 Missense Mutation AGC,GGC S211G NP_001186506.1
NM_001303630.1 412 Missense Mutation AGC,GGC S83G NP_001290559.1
Gene
GIMAP5
Gene Name
GTPase, IMAP family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018384.4 412 Missense Mutation AGC,GGC S7G NP_060854.2

View Full Product Details