Product Details
- SNP ID
-
rs201394816
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:97854583 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATAGCCTCTAAAAATATTACCCA[C/T]GGGCAGCAGTAGTTCGATCTGCGCG
- Phenotype
-
MIM: 108370
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ASNS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1049677] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ASNS
- Gene Name
- asparagine synthetase (glutamine-hydrolyzing)
View Full Product Details