Product Details

SNP ID
rs201600508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151436191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTGTACTCACGCTCCGTCCCC[A/G]TACACCTTGATGGTGTTCACACAGT
Phenotype
MIM: 609603
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRYGN PubMed Links

Gene Details

Gene
CRYGN
Gene Name
crystallin gamma N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308292.1 1037 Intron NP_001295221.1
NM_144727.2 1037 Silent Mutation TAC,TAT Y135Y NP_653328.1
XM_005249952.3 1037 Silent Mutation TAC,TAT Y135Y XP_005250009.1
XM_011515836.1 1037 Silent Mutation TAC,TAT Y135Y XP_011514138.1
XM_011515837.2 1037 Silent Mutation TAC,TAT Y100Y XP_011514139.1
XM_017011773.1 1037 Intron XP_016867262.1
Gene
MIR3907
Gene Name
microRNA 3907
There are no transcripts associated with this gene.

View Full Product Details