Product Details

SNP ID
rs201768315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:81702646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCATTTTGCATAATATGCTACTC[A/G]GACAAAAATACCAGGACGATTTGGA
Phenotype
MIM: 142409
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HGF PubMed Links

Gene Details

Gene
HGF
Gene Name
hepatocyte growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000601.5 2349 Nonsense Mutation CGA,TGA R708* NP_000592.3
NM_001010931.2 2349 Intron NP_001010931.1
NM_001010932.2 2349 Nonsense Mutation CGA,TGA R703* NP_001010932.1
NM_001010933.2 2349 Intron NP_001010933.1
NM_001010934.2 2349 Intron NP_001010934.1
XM_006715956.2 2349 Nonsense Mutation CGA,TGA R708* XP_006716019.1
XM_011516115.2 2349 Nonsense Mutation CGA,TGA R703* XP_011514417.1
XM_017012097.1 2349 Intron XP_016867586.1
XM_017012098.1 2349 Intron XP_016867587.1

View Full Product Details