Product Details

SNP ID
rs202141779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6463120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCGAGCACCCTGAAGGACAGAT[A/G]CTGGTGATGGTCTTTGGCACTTATG
Phenotype
MIM: 609024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDELR2 PubMed Links

Gene Details

Gene
KDELR2
Gene Name
KDEL endoplasmic reticulum protein retention receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100603.1 568 Missense Mutation GCA,GTA A136V NP_001094073.1
NM_006854.3 568 UTR 3 NP_006845.1

View Full Product Details