Product Details

SNP ID
rs201133783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149776417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCCCGTCAGGAGGACCTGGTAC[C/T]CTGTGCCAGCCTCGACCATTACAGC
Phenotype
MIM: 614040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SSPO PubMed Links

Gene Details

Gene
SSPO
Gene Name
SCO-spondin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198455.2 122 Missense Mutation CCC,CTC P41L NP_940857.2
Gene
ZNF467
Gene Name
zinc finger protein 467
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005249959.4 122 Intron XP_005250016.1
XM_005249960.4 122 Intron XP_005250017.1
XM_005249961.4 122 Intron XP_005250018.1
XM_006715864.3 122 Intron XP_006715927.1
XM_011515855.2 122 Intron XP_011514157.1
XM_011515856.2 122 Intron XP_011514158.1
XM_011515857.2 122 Intron XP_011514159.1
XM_011515858.1 122 Intron XP_011514160.1
XM_017011799.1 122 Intron XP_016867288.1

View Full Product Details