Product Details

SNP ID
rs202202571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139134690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCAAACCTGCTGTAGGCAGAGG[C/T]GTACAGCACACTGACAAAAGAAAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTC26 PubMed Links

Gene Details

Gene
TTC26
Gene Name
tetratricopeptide repeat domain 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144920.2 153 Silent Mutation GGC,GGT G13G NP_001138392.1
NM_001144923.2 153 Silent Mutation GGC,GGT G13G NP_001138395.1
NM_001287512.1 153 Silent Mutation GGC,GGT G13G NP_001274441.1
NM_001287513.1 153 UTR 5 NP_001274442.1
NM_001318333.1 153 UTR 5 NP_001305262.1
NM_001321740.1 153 Silent Mutation GGC,GGT G13G NP_001308669.1
NM_001321741.1 153 Silent Mutation GGC,GGT G13G NP_001308670.1
NM_001321742.1 153 Silent Mutation GGC,GGT G13G NP_001308671.1
NM_024926.3 153 Silent Mutation GGC,GGT G13G NP_079202.2

View Full Product Details