Product Details

SNP ID
rs201207694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92213206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGAAAATCTACCTTAGTTTCCA[C/T]GTTGAGGAGATGAAGTCCTTTTATA
Phenotype
MIM: 604214
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRIT1 PubMed Links

Gene Details

Gene
KRIT1
Gene Name
KRIT1, ankyrin repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013406.1 2753 Missense Mutation NP_001013424.1
NM_004912.3 2753 Missense Mutation NP_004903.2
NM_194454.1 2753 Missense Mutation NP_919436.1
NM_194455.1 2753 Missense Mutation NP_919437.1
NM_194456.1 2753 Missense Mutation NP_919438.1
XM_005250660.3 2753 Missense Mutation XP_005250717.1
XM_005250662.3 2753 Missense Mutation XP_005250719.1
XM_005250665.3 2753 Missense Mutation XP_005250722.1
XM_005250666.3 2753 Missense Mutation XP_005250723.1
XM_005250667.2 2753 Missense Mutation XP_005250724.1
XM_005250668.3 2753 Missense Mutation XP_005250725.1
XM_005250669.3 2753 Missense Mutation XP_005250726.1
XM_006716161.3 2753 Missense Mutation XP_006716224.1
XM_006716162.3 2753 Missense Mutation XP_006716225.1
XM_006716163.3 2753 Missense Mutation XP_006716226.1
XM_011516651.2 2753 Missense Mutation XP_011514953.1
XM_011516653.2 2753 Missense Mutation XP_011514955.1
XM_011516654.2 2753 Missense Mutation XP_011514956.1
XM_011516655.2 2753 Missense Mutation XP_011514957.1
XM_011516656.2 2753 Missense Mutation XP_011514958.1
XM_011516657.2 2753 Missense Mutation XP_011514959.1
XM_011516658.2 2753 Missense Mutation XP_011514960.1
XM_011516659.2 2753 Missense Mutation XP_011514961.1
XM_011516660.2 2753 Missense Mutation XP_011514962.1
XM_011516661.2 2753 Missense Mutation XP_011514963.1
XM_017012755.1 2753 Missense Mutation XP_016868244.1
XM_017012756.1 2753 Missense Mutation XP_016868245.1
XM_017012757.1 2753 Missense Mutation XP_016868246.1
XM_017012758.1 2753 Missense Mutation XP_016868247.1
XM_017012759.1 2753 Missense Mutation XP_016868248.1
Gene
MIR1285-1
Gene Name
microRNA 1285-1
There are no transcripts associated with this gene.

View Full Product Details