Product Details
- SNP ID
-
rs201584057
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:99512456 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGTTTTGGTTGTGGTTGTTAGATT[G/T]TTGCCTGCCCTGATGTGCTTCCTCG
- Phenotype
-
MIM: 611272
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZKSCAN5
PubMed Links
Gene Details
- Gene
- ZKSCAN5
- Gene Name
- zinc finger with KRAB and SCAN domains 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318082.1 |
548 |
Missense Mutation |
GTT,TTT |
V140F |
NP_001305011.1 |
NM_001318083.1 |
548 |
Missense Mutation |
GTT,TTT |
V140F |
NP_001305012.1 |
NM_001318084.1 |
548 |
UTR 5 |
|
|
NP_001305013.1 |
NM_014569.3 |
548 |
Missense Mutation |
GTT,TTT |
V140F |
NP_055384.1 |
NM_145102.3 |
548 |
Missense Mutation |
GTT,TTT |
V140F |
NP_659570.1 |
XM_011515999.2 |
548 |
UTR 5 |
|
|
XP_011514301.1 |
XM_017011918.1 |
548 |
Missense Mutation |
GTT,TTT |
V140F |
XP_016867407.1 |
XM_017011919.1 |
548 |
Missense Mutation |
GTT,TTT |
V140F |
XP_016867408.1 |
XM_017011920.1 |
548 |
Missense Mutation |
GTT,TTT |
V140F |
XP_016867409.1 |
XM_017011921.1 |
548 |
Missense Mutation |
GTT,TTT |
V140F |
XP_016867410.1 |
XM_017011922.1 |
548 |
Missense Mutation |
GTT,TTT |
V140F |
XP_016867411.1 |
XM_017011923.1 |
548 |
Missense Mutation |
GTT,TTT |
V140F |
XP_016867412.1 |
XM_017011924.1 |
548 |
Missense Mutation |
GTT,TTT |
V140F |
XP_016867413.1 |
XM_017011925.1 |
548 |
UTR 5 |
|
|
XP_016867414.1 |
XM_017011926.1 |
548 |
UTR 5 |
|
|
XP_016867415.1 |
XM_017011927.1 |
548 |
UTR 5 |
|
|
XP_016867416.1 |
View Full Product Details