Product Details

SNP ID
rs201209040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:55978223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAGAAAAGATGCCCACTCCAAT[C/G]TCCTAGCCAAAAAGGAAACAAGCAA
Phenotype
MIM: 603004
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GBAS PubMed Links

Gene Details

Gene
GBAS
Gene Name
glioblastoma amplified sequence
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202469.1 223 Missense Mutation CTC,GTC L64V NP_001189398.1
NM_001483.2 223 Missense Mutation CTC,GTC L64V NP_001474.1

View Full Product Details