Product Details
- SNP ID
-
rs201322412
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:17794179 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGGAAATAAGGTTTCTAAAAAGC[C/T]TTCCAAGAAGACATAAACCATTCTC
- Phenotype
-
MIM: 606589
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNX13
PubMed Links
Gene Details
- Gene
- SNX13
- Gene Name
- sorting nexin 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015132.4 |
2884 |
Missense Mutation |
AGC,GGC |
S914G |
NP_055947.1 |
XM_005249672.1 |
2884 |
Missense Mutation |
AGC,GGC |
S925G |
XP_005249729.1 |
XM_005249673.4 |
2884 |
Intron |
|
|
XP_005249730.1 |
XM_011515229.1 |
2884 |
Missense Mutation |
AGC,GGC |
S871G |
XP_011513531.1 |
XM_011515230.2 |
2884 |
Missense Mutation |
AGC,GGC |
S845G |
XP_011513532.1 |
XM_011515231.2 |
2884 |
Missense Mutation |
AGC,GGC |
S845G |
XP_011513533.1 |
XM_011515233.2 |
2884 |
Missense Mutation |
AGC,GGC |
S823G |
XP_011513535.1 |
XM_017011893.1 |
2884 |
Missense Mutation |
AGC,GGC |
S860G |
XP_016867382.1 |
XM_017011894.1 |
2884 |
Missense Mutation |
AGC,GGC |
S845G |
XP_016867383.1 |
XM_017011895.1 |
2884 |
Missense Mutation |
AGC,GGC |
S812G |
XP_016867384.1 |
XM_017011896.1 |
2884 |
Missense Mutation |
AGC,GGC |
S796G |
XP_016867385.1 |
XM_017011897.1 |
2884 |
Missense Mutation |
AGC,GGC |
S962G |
XP_016867386.1 |
View Full Product Details