Product Details

SNP ID
rs201436002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:195589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTGAAGGTGCAGAATTCCTCT[A/C]CCCCGGGGAGGCGGCCGTGGACTCC
Phenotype
MIM: 611061
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM20C PubMed Links

Gene Details

Gene
FAM20C
Gene Name
family with sequence similarity 20 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020223.3 882 Missense Mutation TAC,TCC Y214S NP_064608.2
XM_017012450.1 882 Missense Mutation TAC,TCC Y214S XP_016867939.1
XM_017012451.1 882 Missense Mutation TAC,TCC Y214S XP_016867940.1
XM_017012452.1 882 Missense Mutation TAC,TCC Y214S XP_016867941.1
XM_017012453.1 882 Missense Mutation TAC,TCC Y214S XP_016867942.1
XM_017012454.1 882 Missense Mutation TAC,TCC Y214S XP_016867943.1
XM_017012455.1 882 Intron XP_016867944.1

View Full Product Details