Product Details

SNP ID
rs202032471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100707175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTGATGAGCTGGAGCCAGAGAC[C/T]GACACACGGGAGCCACTGACTCGGA
Phenotype
MIM: 606113
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POP7 PubMed Links

Gene Details

Gene
POP7
Gene Name
POP7 homolog, ribonuclease P/MRP subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005837.2 607 Silent Mutation ACC,ACT T115T NP_005828.2

View Full Product Details