Product Details

SNP ID
rs202145036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:47988808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACCTGGAGTTCAAATGTTTGAA[C/T]GGTGGTTCCTGTTTTGTTATATATA
Phenotype
MIM: 603760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HUS1 PubMed Links

Gene Details

Gene
HUS1
Gene Name
HUS1 checkpoint clamp component
There are no transcripts associated with this gene.

Gene
SUN3
Gene Name
Sad1 and UNC84 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030019.1 1021 Missense Mutation ATT,GTT I312V NP_001025190.1
NM_001284350.1 1021 Missense Mutation ATT,GTT I300V NP_001271279.1
NM_152782.3 1021 Missense Mutation ATT,GTT I312V NP_689995.3
XM_017011930.1 1021 Missense Mutation ATT,GTT I312V XP_016867419.1

View Full Product Details