Product Details
- SNP ID
-
rs201698982
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149720944 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGCGCTATGGACAGCCCCGAGAG[C/T]GAGAGCCGGGGAGCCAGCCTGGATG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRBA1
PubMed Links
Gene Details
- Gene
- KRBA1
- Gene Name
- KRAB-A domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001290187.1 |
589 |
Silent Mutation |
AGC,AGT |
S123S |
NP_001277116.1 |
NM_032534.3 |
589 |
Silent Mutation |
AGC,AGT |
S88S |
NP_115923.2 |
XM_011516636.2 |
589 |
Silent Mutation |
AGC,AGT |
S160S |
XP_011514938.1 |
XM_011516637.2 |
589 |
Silent Mutation |
AGC,AGT |
S160S |
XP_011514939.1 |
XM_011516638.2 |
589 |
Silent Mutation |
AGC,AGT |
S145S |
XP_011514940.1 |
XM_011516639.2 |
589 |
Silent Mutation |
AGC,AGT |
S160S |
XP_011514941.1 |
XM_011516640.2 |
589 |
Silent Mutation |
AGC,AGT |
S160S |
XP_011514942.1 |
XM_011516641.2 |
589 |
Silent Mutation |
AGC,AGT |
S123S |
XP_011514943.1 |
XM_011516642.2 |
589 |
Silent Mutation |
AGC,AGT |
S88S |
XP_011514944.1 |
XM_011516643.2 |
589 |
Silent Mutation |
AGC,AGT |
S88S |
XP_011514945.1 |
View Full Product Details