Product Details
- SNP ID
-
rs202071453
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:99560686 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGGACACCGACATGGAACAGGGA[C/G]TCACTGGGGGTAAGGCAGAGAAAGC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FAM200A
PubMed Links
Gene Details
- Gene
- FAM200A
- Gene Name
- family with sequence similarity 200 member A
There are no transcripts associated with this gene.
- Gene
- ZNF655
- Gene Name
- zinc finger protein 655
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001009958.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
NP_001009958.1 |
NM_001009960.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
NP_001009960.1 |
NM_001083956.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
NP_001077425.1 |
NM_001085366.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
NP_001078835.1 |
NM_001085367.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
NP_001078836.1 |
NM_001085368.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
NP_001078837.1 |
NM_024061.3 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
NP_076966.1 |
NM_138494.2 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
NP_612503.1 |
XM_017012598.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868087.1 |
XM_017012599.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868088.1 |
XM_017012600.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868089.1 |
XM_017012601.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868090.1 |
XM_017012602.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868091.1 |
XM_017012603.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868092.1 |
XM_017012604.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868093.1 |
XM_017012605.1 |
364 |
UTR 5 |
|
|
XP_016868094.1 |
XM_017012606.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868095.1 |
XM_017012607.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868096.1 |
XM_017012608.1 |
364 |
UTR 5 |
|
|
XP_016868097.1 |
XM_017012609.1 |
364 |
UTR 5 |
|
|
XP_016868098.1 |
XM_017012610.1 |
364 |
UTR 5 |
|
|
XP_016868099.1 |
XM_017012611.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868100.1 |
XM_017012612.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868101.1 |
XM_017012613.1 |
364 |
Missense Mutation |
CTC,GTC |
L43V |
XP_016868102.1 |
View Full Product Details