Product Details

SNP ID
rs201267113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101616237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTTACACATCGGCCTTGACGAA[A/G]CCTTTCCCTTCAGTGTCGAACACTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYL10 PubMed Links

Gene Details

Gene
MYL10
Gene Name
myosin light chain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138403.4 414 Silent Mutation GGC,GGT G172G NP_612412.2
XM_017012793.1 414 Silent Mutation GGC,GGT G93G XP_016868282.1

View Full Product Details